Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRIG3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136051.2 | NP_001129523.1 | 18 | 2807 | ||
NM_153377.4 | NP_700356.2 | 18 | 3226 | |||
XM_011537893.2 | XP_011536195.1 | 18 | 2898 | |||
GenBank mRNA | AK074921.1 | - | 9 | 1726 | ||
AK130083.1 | - | 4 | 694 | |||
AK292857.1 | - | 18 | 3207 | |||
AL833710.1 | - | 7 | 1563 | |||
AY358288.1 | - | 18 | 3169 | |||
AY358295.1 | - | 18 | 2807 | |||
AY505340.1 | - | 18 | 2977 | |||
BC126169.1 | - | 18 | 3122 | 8991885 | ||
BC126171.1 | - | 18 | 3122 | 8991887 | ||
BC143960.1 | - | 19 | 3232 |