Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C12orf54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152319.3 | NP_689532.1 | 6, 7 | 316 | ||
XM_005268636.3 | XP_005268693.1 | 5, 6 | 198 | |||
XM_011537896.2 | XP_011536198.1 | 8, 9 | 964 | |||
XM_017018796.1 | XP_016874285.1 | 10, 11 | 1154 | |||
GenBank mRNA | AY255793.1 | - | 5, 6 | 221 | ||
BC031670.2 | - | 5, 6 | 242 | 5165130 | ||
BQ429029.1 | - | 6, 7 | 319 |