Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BTBD11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017523.1 | NP_001017523.1 | 13, 14 | 1659 | ||
NM_001018072.1 | NP_001018082.1 | 15, 16 | 3519 | |||
XM_005268645.3 | XP_005268702.1 | 15, 16 | 1887 | |||
XM_011537909.2 | XP_011536211.1 | 15, 16 | 2442 | |||
XM_011537910.2 | XP_011536212.1 | 15, 16 | 2396 | |||
XM_017018807.1 | XP_016874296.1 | 14, 15 | 4669 | |||
GenBank mRNA | AK091276.1 | - | 5, 6 | 494 | ||
AK122818.1 | - | 15, 16 | 1956 | |||
AK127012.1 | - | 12, 13 | 1429 | |||
AK127295.1 | - | 13, 14 | 1659 | |||
AK128445.1 | - | 7, 8 | 898 | |||
AY373588.1 | - | 11, 12 | 1353 | |||
BC093627.1 | - | 4, 5 | 325 | |||
BC093629.1 | - | 4, 5 | 325 | |||
BC101561.1 | - | 4, 5 | 325 | |||
BC101563.1 | - | 4, 5 | 325 |