Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALKBH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001655.2 | NP_001001655.1 | 4 | 819 | ||
NM_001145374.1 | NP_001138846.1 | 4 | 969 | |||
NM_001145375.1 | NP_001138847.1 | 4 | 716 | |||
NM_001205179.1 | NP_001192108.1 | 3 | 667 | |||
NM_001205180.1 | NP_001192109.1 | 3 | 620 | |||
XM_005253835.4 | XP_005253892.1 | 4 | 985 | |||
XM_005253836.1 | XP_005253893.1 | 3 | 913 | |||
GenBank mRNA | AB277859.1 | - | 2 | 383 | ||
AI278638.1 | - | 1 | 234 | |||
AY754389.1 | - | 4 | 804 | |||
BC070489.1 | - | 4 | 771 | 6048174 | ||
BU623121.1 | - | 1 | 243 |