Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPPL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139015.4 | NP_620584.2 | 3, 4 | 674 | ||
XM_011537925.2 | XP_011536227.1 | 3, 4 | 204 | |||
GenBank mRNA | AB252457.1 | - | 3, 4 | 266 | ||
AJ345030.1 | - | 3, 4 | 242 | |||
AK074916.1 | - | 2, 3 | 157 | |||
AK223590.1 | - | 3, 4 | 670 | |||
AK291161.1 | - | 3, 4 | 496 | |||
AY169313.1 | - | 3, 4 | 448 | |||
BC073910.1 | - | 3, 4 | 455 | 4538522 | ||
BC101625.1 | - | 3, 4 | 269 | 8069131 | ||
BC101627.1 | - | 3, 4 | 269 | 8069133 | ||
BU932077.1 | - | 3, 4 | 674 |