Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRAPPC6B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079537.1 | NP_001073005.1 | 2 | 433 | ||
NM_177452.3 | NP_803235.1 | 2 | 433 | |||
XM_011536409.2 | XP_011534711.1 | 3 | 453 | |||
XM_017020964.1 | XP_016876453.1 | 2 | 406 | |||
GenBank mRNA | AK056690.1 | - | 2 | 403 | ||
AK310730.1 | - | 2 | 210 | |||
AK315900.1 | - | 2 | 328 | |||
AL833179.1 | - | 2 | 317 | |||
BC047328.1 | - | 2 | 317 | 5534151 | ||
BX161510.1 | - | 2 | 325 | |||
BX248013.1 | - | 2 | 356 |