Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SOCS4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080867.2 | NP_543143.1 | 2 | 619 | ||
NM_199421.1 | NP_955453.1 | 3 | 748 | |||
XM_011536425.1 | XP_011534727.1 | 3 | 802 | |||
XM_011536426.1 | XP_011534728.1 | 3 | 717 | |||
GenBank mRNA | AF424815.1 | - | 1 | 399 | ||
AK124083.1 | - | 2 | 472 | |||
BC060790.1 | - | 3 | 626 | 30338646 | ||
BX161418.1 | - | 2 | 515 | |||
BX648734.1 | - | 2 | 405 | |||
CD109883.1 | - | 2 | 646 | |||
CR933659.1 | - | 2 | 509 |