Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C15orf40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144597.2 | NP_653198.2 | 4 | 445 | ||
GenBank mRNA | AK125586.1 | - | 3 | 1531 | ||
AK312160.1 | - | 4 | 419 | |||
AL602775.1 | - | 2 | 190 | |||
BC019820.1 | - | 4 | 410 | 4449390 | ||
KU178855.1 | - | 4 | 330 | |||
KU178856.1 | - | 3 | 202 |