Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SENP8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166340.1 | NP_001159812.1 | 2 | 350 | ||
NM_001172109.1 | NP_001165580.1 | 3 | 504 | |||
NM_001172110.1 | NP_001165581.1 | 3 | 346 | |||
NM_001172111.1 | NP_001165582.1 | 3 | 363 | |||
NM_145204.3 | NP_660205.3 | 2 | 239 | |||
XM_005254157.3 | XP_005254214.1 | 2 | 119 | |||
XM_011521215.2 | XP_011519517.1 | 3 | 637 | |||
XM_011521217.2 | XP_011519519.1 | 2 | 227 | |||
GenBank mRNA | AF308450.1 | - | 2 | 143 | ||
AK097557.1 | - | 2 | 350 | |||
AK292298.1 | - | 2 | 241 | |||
AY008293.1 | - | 2 | 203 | |||
BC031411.1 | - | 2 | 116 | 5195120 | ||
BP370384.1 | - | 3 | 346 | |||
CN367895.1 | - | 3 | 330 | |||
DB033241.1 | - | 2 | 241 | |||
DB048641.1 | - | 3 | 504 |