Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRC28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284400.2 | NP_001271329.1 | 5 | 529 | ||
NM_001321675.1 | NP_001308604.1 | 5 | 525 | |||
NM_001321676.1 | NP_001308605.1 | 5 | 529 | |||
NM_001321677.1 | NP_001308606.1 | 5 | 529 | |||
NM_001321678.1 | NP_001308607.1 | 5 | 377 | |||
NM_001321679.1 | NP_001308608.1 | 4 | 488 | |||
NM_001321680.1 | NP_001308609.1 | 4 | 484 | |||
NM_144598.4 | NP_653199.2 | 5 | 529 | |||
NR_135753.1 | 5 | 529 | ||||
NR_135754.1 | 5 | 377 | ||||
NR_135755.1 | 4 | 484 | ||||
NR_135756.1 | 4 | 488 | ||||
NR_135757.1 | 5 | 529 | ||||
NR_135758.1 | 4 | 488 | ||||
NR_135759.1 | 4 | 484 | ||||
XM_006720389.3 | XP_006720452.1 | 5 | 529 | |||
XM_011521218.2 | XP_011519520.1 | 5 | 565 | |||
XM_011521220.1 | XP_011519522.1 | 5 | 529 | |||
XM_011521221.2 | XP_011519523.1 | 5 | 529 | |||
XM_017021914.1 | XP_016877403.1 | 5 | 813 | |||
GenBank mRNA | AK091588.1 | - | 4 | 391 | ||
AK292887.1 | - | 5 | 386 | |||
AK293832.1 | - | 5 | 386 | |||
AK295161.1 | - | 5 | 404 | |||
AK296352.1 | - | 5 | 431 | |||
AY358082.1 | - | 5 | 386 | |||
BC047306.1 | - | 5 | 444 | 4999965 |