Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NTAN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270766.1 | NP_001257695.1 | 4, 5 | 429 | ||
NM_001270767.1 | NP_001257696.1 | 3, 4 | 363 | |||
NM_173474.3 | NP_775745.1 | 5, 6 | 532 | |||
XM_011522355.2 | XP_011520657.1 | 2, 3 | 367 | |||
GenBank mRNA | AF092440.1 | - | 5, 6 | 439 | ||
AK092915.1 | - | 1, 2 | 1715 | |||
BC017336.2 | - | 5, 6 | 429 | 4872717 | ||
BF939373.1 | - | 3, 4 | 270 | |||
DC383993.1 | - | 4, 5 | 377 |