Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NTAN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173474.3 | NP_775745.1 | 2 | 207 | ||
GenBank mRNA | AF092440.1 | - | 2 | 114 | ||
BC017336.2 | - | 2 | 104 | 4872717 | ||
HY036316.1 | - | 2 | 208 |