Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IQCK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001305121.1 | NP_001292050.1 | 3 | 978 | ||
NM_001305122.1 | NP_001292051.1 | 4 | 1043 | |||
NM_153208.2 | NP_694940.1 | 4 | 1043 | |||
NR_130967.1 | 4 | 1043 | ||||
NR_130968.1 | 4 | 1043 | ||||
XM_011545736.2 | XP_011544038.1 | 3 | 378 | |||
XM_017022933.1 | XP_016878422.1 | 3 | 1089 | |||
GenBank mRNA | AF520569.1 | - | 4 | 1043 | ||
AF520570.1 | - | 4 | 1043 | |||
AK000122.1 | - | 3 | 978 | |||
AK093894.1 | - | 3 | 314 | |||
AK301933.1 | - | 3 | 366 | |||
AK303998.1 | - | 2 | 271 | |||
AK311216.1 | - | 3 | 336 | |||
AK315672.1 | - | 3 | 366 | |||
BC034823.1 | - | 3 | 374 | 5165639 |