Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: USH1G | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282489.2 | NP_001269418.1 | 2 | 1088 | ||
NM_173477.4 | NP_775748.2 | 2 | 1141 | |||
XM_011524296.2 | XP_011522598.1 | 2 | 1114 | |||
GenBank mRNA | AK091243.1 | - | 2 | 1134 | ||
AK289804.1 | - | 2 | 1138 | |||
AK296899.1 | - | 2 | 1081 | |||
BC101096.2 | - | 2 | 1028 | 40010028 | ||
BC101097.1 | - | 2 | 1028 | 40010033 | ||
BC101098.2 | - | 2 | 1028 | 40010036 | ||
BC101099.1 | - | 2 | 1028 | 40010038 |