Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD300LB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_174892.3 | NP_777552.3 | 2 | 226 | ||
XM_005257027.3 | XP_005257084.1 | 2 | 226 | |||
GenBank mRNA | AF427618.1 | - | 2 | 126 | ||
AK290925.1 | - | 2 | 226 | |||
AK298665.1 | - | 2 | 227 | |||
AY359025.1 | - | 2 | 155 | |||
AY646929.1 | - | 2 | 173 | |||
BC028091.1 | - | 2 | 218 | 5219239 | ||
DA673717.1 | - | 2 | 228 |