Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNTD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173478.2 | NP_775749.2 | 5 | 887 | ||
XM_005257043.4 | XP_005257100.1 | 5 | 650 | |||
XM_005257044.4 | XP_005257101.1 | 5 | 668 | |||
XM_005257045.4 | XP_005257102.1 | 4 | 810 | |||
XM_006721690.2 | XP_006721753.1 | 5 | 887 | |||
XM_011524310.2 | XP_011522612.1 | 5 | 834 | |||
XM_011524311.2 | XP_011522613.1 | 4 | 492 | |||
XM_017024171.1 | XP_016879660.1 | 4 | 810 | |||
GenBank mRNA | AK097456.1 | - | 5 | 803 | ||
AK302094.1 | - | 5 | 620 | |||
AL833052.1 | - | 3 | 267 | |||
BC026187.2 | - | 5 | 931 | 5267540 |