Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEZ6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098635.1 | NP_001092105.1 | 15, 16 | 3041 | ||
NM_001290202.1 | NP_001277131.1 | 15, 16 | 2877 | |||
NM_178860.4 | NP_849191.3 | 15, 16 | 3041 | |||
GenBank mRNA | AF502129.1 | - | 15, 16 | 3129 | ||
AK054913.1 | - | 6, 7 | 902 | |||
AK055383.1 | - | 9, 10 | 1313 | |||
AK091522.1 | - | 15, 16 | 2877 | |||
AK096473.1 | - | 14, 15 | 2290 | |||
AK125377.1 | - | 15, 16 | 3002 | |||
AK223620.1 | - | 15, 16 | 3003 | |||
AL834405.1 | - | 14, 15 | 2786 | |||
AY038048.1 | - | 15, 16 | 3032 |