Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC43A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284498.1 | NP_001271427.1 | 3 | 596 | ||
NM_001321364.1 | NP_001308293.1 | 3 | 394 | |||
NM_001321365.1 | NP_001308294.1 | 3 | 563 | |||
NM_152346.2 | NP_689559.1 | 3 | 394 | |||
XM_017024177.1 | XP_016879666.1 | 3 | 571 | |||
XM_017024178.1 | XP_016879667.1 | 3 | 604 | |||
GenBank mRNA | AB120364.1 | - | 3 | 562 | ||
AF289592.1 | - | 3 | 439 | |||
BC027923.1 | - | 3 | 344 | 5309751 | ||
BC071859.1 | - | 3 | 394 | 6572285 |