Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EFCAB13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195192.1 | NP_001182121.1 | 10 | 1203 | ||
NM_152347.4 | NP_689560.3 | 12 | 1491 | |||
GenBank mRNA | AK097661.1 | - | 9 | 1050 | ||
BC036407.2 | - | 12 | 1471 | 5266195 | ||
BC037876.1 | - | 10 | 1143 | 5273577 |