Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR81 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163673.1 | NP_001157145.1 | 6, 7 | 936 | ||
NM_001163809.1 | NP_001157281.1 | 6, 7 | 4621 | |||
NM_001163811.1 | NP_001157283.1 | 6, 7 | 1072 | |||
NM_152348.3 | NP_689561.2 | 7, 8 | 1573 | |||
XM_011523651.2 | XP_011521953.1 | 7, 8 | 1558 | |||
XM_017024184.1 | XP_016879673.1 | 6, 7 | 4644 | |||
GenBank mRNA | AK074111.1 | - | 6, 7 | 1875 | ||
AK074356.1 | - | 1, 2 | 164 | |||
AK091136.1 | - | 7, 8 | 1557 | |||
AK122972.1 | - | 6, 7 | 1883 | |||
AK123896.1 | - | 6, 7 | 1063 | |||
AK127946.1 | - | 6, 7 | 1072 | |||
AK298567.1 | - | 6, 7 | 936 | |||
AK300986.1 | - | 4, 5 | 553 | |||
AL834379.1 | - | 6, 7 | 1397 | |||
BC092513.1 | - | 6, 7 | 1348 | 3092839 | ||
BC114519.1 | - | 5, 6 | 831 | 40036610 | ||
BC114568.1 | - | 6, 7 | 898 | 40036619 |