Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM210A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098801.1 | NP_001092271.1 | 3 | 445 | ||
NM_152352.3 | NP_689565.2 | 2 | 305 | |||
GenBank mRNA | AK055618.1 | - | 3 | 401 | ||
DA560663.1 | - | 3 | 375 | |||
DA895646.1 | - | 2 | 275 | |||
DA984075.1 | - | 2 | 302 |