Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLDND2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152353.2 | NP_689566.1 | 3 | 791 | ||
XM_011526425.2 | XP_011524727.1 | 3 | 1043 | |||
XM_011526428.2 | XP_011524730.1 | 3 | 1090 | |||
XM_017026244.1 | XP_016881733.1 | 2 | 1472 | |||
XM_017026245.1 | XP_016881734.1 | 3 | 1112 | |||
XM_017026246.1 | XP_016881735.1 | 2 | 1352 | |||
XM_017026247.1 | XP_016881736.1 | 2 | 1472 | |||
GenBank mRNA | BC029518.1 | - | 3 | 791 | 5295481 |