Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CALR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145046.4 | NP_659483.2 | 7 | 915 | ||
GenBank mRNA | AB576176.1 | - | 7 | 852 | ||
AK058084.1 | - | 7 | 907 | |||
BC014595.2 | - | 7 | 915 | 4822010 |