Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPT1C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136052.2 | NP_001129524.1 | 18 | 2351 | ||
NM_001199752.2 | NP_001186681.1 | 18 | 2448 | |||
NM_001199753.1 | NP_001186682.1 | 18 | 2384 | |||
NM_152359.2 | NP_689572.1 | 17 | 2315 | |||
NR_108072.1 | 19 | 2546 | ||||
XM_005258505.2 | XP_005258562.1 | 18 | 2421 | |||
XM_005258506.4 | XP_005258563.1 | 17 | 2337 | |||
XM_006723009.3 | XP_006723072.1 | 15 | 2153 | |||
XM_011526438.2 | XP_011524740.1 | 18 | 2477 | |||
XM_011526439.2 | XP_011524741.1 | 18 | 2541 | |||
XM_011526440.2 | XP_011524742.1 | 17 | 2258 | |||
XM_017026265.1 | XP_016881754.1 | 17 | 2305 | |||
XM_017026266.1 | XP_016881755.1 | 17 | 2226 | |||
XM_017026267.1 | XP_016881756.1 | 17 | 2211 | |||
XM_017026268.1 | XP_016881757.1 | 16 | 2054 | |||
XM_017026269.1 | XP_016881758.1 | 16 | 2164 | |||
XM_017026270.1 | XP_016881759.1 | 16 | 1932 | |||
XM_017026271.1 | XP_016881760.1 | 16 | 2021 | |||
XM_017026272.1 | XP_016881761.1 | 16 | 2113 | |||
XM_017026273.1 | XP_016881762.1 | 16 | 2098 | |||
GenBank mRNA | AF357970.1 | - | 17 | 2273 | ||
AK074389.1 | - | 10 | 1294 | |||
AK096516.1 | - | 19 | 2542 | |||
AK289713.1 | - | 18 | 2440 | |||
AK290092.1 | - | 18 | 2356 | |||
BC029104.1 | - | 18 | 2315 | 3872727 |