Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IRGQ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007561.2 | NP_001007562.1 | 3 | 1348 | ||
XM_005258515.3 | XP_005258572.1 | 3 | 1427 | |||
GenBank mRNA | AF322648.1 | - | 2 | 1378 | ||
BC137020.1 | - | 2 | 1310 | 9021013 | ||
BC137021.1 | - | 2 | 1310 | 9021014 | ||
BC144451.1 | - | 2 | 1310 | 9052977 |