Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MISP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173481.3 | NP_775752.1 | 2 | 1740 | ||
XM_011527685.2 | XP_011525987.1 | 2 | 1785 | |||
XM_011527686.2 | XP_011525988.1 | 2 | 1878 | |||
GenBank mRNA | BC042125.2 | - | 2 | 1707 | 3848272 | |
BC052236.1 | - | 2 | 1727 | 5431360 | ||
BX648389.1 | - | 2 | 1721 | |||
BX648815.1 | - | 1 | 928 |