Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBXO27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178820.4 | NP_849142.1 | 5 | 802 | ||
XM_017026290.1 | XP_016881779.1 | 5 | 761 | |||
GenBank mRNA | AF436061.1 | - | 5 | 745 | ||
BC014527.2 | - | 5 | 764 | 3841901 | ||
BC030060.1 | - | 5 | 731 | 5421414 | ||
BX377760.2 | - | 5 | 802 |