Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TDRD10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098475.1 | NP_001091945.1 | 11 | 1675 | ||
NM_182499.3 | NP_872305.3 | 11 | 1675 | |||
XM_006711156.2 | XP_006711219.1 | 6 | 1305 | |||
XM_011509152.1 | XP_011507454.1 | 12 | 1064 | |||
XM_011509153.1 | XP_011507455.1 | 12 | 1064 | |||
XM_011509154.1 | XP_011507456.1 | 12 | 1022 | |||
XM_011509155.1 | XP_011507457.1 | 12 | 1064 | |||
XM_011509156.1 | XP_011507458.1 | 11 | 989 | |||
XM_011509157.1 | XP_011507459.1 | 11 | 989 | |||
XM_011509158.2 | XP_011507460.1 | 7 | 779 | |||
XM_011509159.2 | XP_011507461.1 | 7 | 770 | |||
XM_011509160.2 | XP_011507462.1 | 7 | 815 | |||
XM_011509161.1 | XP_011507463.1 | 7 | 783 | |||
XM_017000255.1 | XP_016855744.1 | 6 | 709 | |||
GenBank mRNA | AK302144.1 | - | 11 | 922 | ||
AL713777.1 | - | 6 | 580 | |||
AY358195.1 | - | 5 | 444 | |||
BC044864.1 | - | 11 | 1674 | 4836747 | ||
BC100895.1 | - | 6 | 553 | |||
BC100896.1 | - | 6 | 553 | |||
BC100897.2 | - | 6 | 553 | |||
BC100898.2 | - | 6 | 553 |