Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR63 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001288563.1 | NP_001275492.1 | 15 | 1800 | ||
NM_145172.4 | NP_660155.2 | 16 | 1917 | |||
GenBank mRNA | AK054629.1 | - | 15 | 1754 | ||
AK292603.1 | - | 16 | 1888 | |||
AY049724.1 | - | 16 | 1898 | |||
BC040265.1 | - | 16 | 1871 | 5165779 | ||
BX648851.1 | - | 16 | 1923 |