Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TYW3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162916.1 | NP_001156388.1 | 2, 3 | 358 | ||
NM_138467.2 | NP_612476.1 | 3, 4 | 453 | |||
NR_027962.1 | 3, 4 | 544 | ||||
XM_006710347.1 | XP_006710410.1 | 3, 4 | 466 | |||
GenBank mRNA | AK098237.1 | - | 3, 4 | 453 | ||
BC009514.1 | - | 3, 4 | 387 | |||
BC071765.1 | - | 3, 4 | 453 | 3931600 |