Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TPRG1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182752.3 | NP_877429.2 | 4 | 591 | ||
GenBank mRNA | AK025464.1 | - | 3 | 311 | ||
AK096427.1 | - | 4 | 568 | |||
AK289919.1 | - | 4 | 541 | |||
AK298343.1 | - | 4 | 430 | |||
BC019034.2 | - | 3 | 356 | 4576259 | ||
BI547609.1 | - | 4 | 585 |