Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYOM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152372.3 | NP_689585.3 | 16 | 2095 | ||
GenBank mRNA | AK125972.1 | - | 14 | 2137 | ||
AK128233.1 | - | 8 | 1155 | |||
BC067101.1 | - | 16 | 2105 | 5928128 | ||
BX537911.1 | - | 15 | 1819 | |||
BX647453.1 | - | 16 | 2056 | |||
BX647455.1 | - | 16 | 2095 | |||
BX647644.1 | - | 16 | 2056 |