Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VWA5B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039500.2 | NP_001034589.2 | 17 | 2877 | ||
XM_006710360.3 | XP_006710423.2 | 14 | 2152 | |||
XM_011540683.2 | XP_011538985.1 | 17 | 2839 | |||
XM_011540684.2 | XP_011538986.1 | 17 | 2964 | |||
XM_011540685.2 | XP_011538987.1 | 17 | 2976 | |||
XM_011540688.2 | XP_011538990.1 | 17 | 2839 | |||
XM_011540690.2 | XP_011538992.1 | 16 | 2662 | |||
XM_011540691.2 | XP_011538993.1 | 16 | 2653 | |||
XM_011540692.2 | XP_011538994.1 | 17 | 2833 | |||
XM_011540693.1 | XP_011538995.1 | 17 | 2839 | |||
XM_011540695.1 | XP_011538997.1 | 17 | 2839 | |||
XM_011540697.2 | XP_011538999.1 | 11 | 2210 | |||
XM_017000305.1 | XP_016855794.1 | 11 | 1956 | |||
XM_017000306.1 | XP_016855795.1 | 12 | 2117 | |||
GenBank mRNA | AK057346.1 | - | 6 | 960 | ||
AK295797.1 | - | 17 | 2861 | |||
BC101378.1 | - | 6 | 957 | |||
BC101379.2 | - | 6 | 957 | |||
BC101380.1 | - | 6 | 957 | |||
BC101381.1 | - | 6 | 957 |