Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LIX1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153713.2 | NP_714924.1 | 2 | 439 | ||
XM_017000316.1 | XP_016855805.1 | 2 | 458 | |||
GenBank mRNA | AK128733.1 | - | 3 | 280 | ||
BC035727.1 | - | 2 | 421 | 5723677 | ||
CR627381.1 | - | 1 | 77 |