Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LIX1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153713.2 | NP_714924.1 | 4 | 764 | ||
GenBank mRNA | AK123675.1 | - | 1 | 16 | ||
AK128733.1 | - | 5 | 605 | |||
BC035727.1 | - | 4 | 746 | 5723677 | ||
CR627381.1 | - | 3 | 402 |