Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IQGAP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178229.4 | NP_839943.2 | 31, 32 | 4041 | ||
XM_011509198.2 | XP_011507500.1 | 31, 32 | 4030 | |||
XM_011509200.2 | XP_011507502.1 | 20, 21 | 3210 | |||
XM_011509201.2 | XP_011507503.1 | 19, 20 | 2599 | |||
XM_017000317.1 | XP_016855806.1 | 31, 32 | 4027 | |||
XM_017000318.1 | XP_016855807.1 | 29, 30 | 3784 | |||
GenBank mRNA | AB105103.1 | - | 31, 32 | 3977 | ||
AK122653.1 | - | 17, 18 | 2249 | |||
AY253300.1 | - | 31, 32 | 3966 | |||
BC131536.1 | - | 31, 32 | 3999 | 40082932 |