Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARHGEF19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153213.3 | NP_694945.2 | 13, 14 | 2210 | ||
XM_011540706.2 | XP_011539008.1 | 14, 15 | 2374 | |||
GenBank mRNA | AL137736.1 | - | 8, 9 | 966 | ||
BC012982.2 | - | 12, 13 | 1786 | |||
BC040640.1 | - | 13, 14 | 2210 | 3847806 |