Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NKAIN4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152864.3 | NP_690603.3 | 2 | 210 | ||
XM_005260192.1 | XP_005260249.1 | 2 | 172 | |||
XM_005260194.3 | XP_005260251.1 | 2 | 631 | |||
XM_011528527.1 | XP_011526829.1 | 2 | 172 | |||
XM_011528528.2 | XP_011526830.1 | 2 | 634 | |||
XM_011528529.2 | XP_011526831.1 | 2 | 620 | |||
XM_017027636.1 | XP_016883125.1 | 2 | 142 | |||
XM_017027637.1 | XP_016883126.1 | 2 | 624 | |||
XM_017027638.1 | XP_016883127.1 | 2 | 630 | |||
XM_017027639.1 | XP_016883128.1 | 2 | 616 | |||
GenBank mRNA | AK126246.1 | - | 2 | 133 | ||
AK225765.1 | - | 2 | 164 | |||
AK308958.1 | - | 2 | 302 | |||
BC041812.1 | - | 2 | 208 | 5302801 | ||
BG708360.1 | - | 2 | 214 |