Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TANGO2 SIRNA Assays ›
Gene Symbol:
TANGO2
Gene Name:
transport and golgi organization 2 homolog
Gene Aliases:
C22orf25, MECRCN
Chromosome Location:
Chr.22: 20017000 - 20067164 on Build GRCh38
UniGene ID:
Hs.474233
Species:
Human
siRNA ID
HSS134162
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TANGO2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001283106.2
NP_001270035.1
9, 10
1059
NM_001283116.2
NP_001270045.1
8, 9
813
NM_001283129.2
NP_001270058.1
8, 9
1053
NM_001283148.2
NP_001270077.1
8, 9
811
NM_001283154.2
NP_001270083.1
8, 9
923
NM_001283179.2
NP_001270108.1
6, 7
754
NM_001283186.2
NP_001270115.1
6, 7
627
NM_001283199.2
NP_001270128.1
6, 7
700
NM_001283235.2
NP_001270164.1
7, 8
698
NM_001283248.2
NP_001270177.1
5, 6
473
NM_001322143.1
NP_001309072.1
8, 9
1126
NM_001322144.1
NP_001309073.1
8, 9
1051
NM_001322145.1
NP_001309074.1
6, 7
940
NM_001322147.1
NP_001309076.1
6, 7
867
NM_001322148.1
NP_001309077.1
8, 9
1130
NM_001322149.1
NP_001309078.1
6, 7
828
NM_001322160.1
NP_001309089.1
8, 9
1013
NM_001322163.1
NP_001309092.1
7, 8
873
NM_001322166.1
NP_001309095.1
6, 7
700
NM_001322167.1
NP_001309096.1
6, 7
752
NM_001322169.1
NP_001309098.1
6, 7
625
NM_001322171.1
NP_001309100.1
7, 8
771
NM_001322172.1
NP_001309101.1
7, 8
1063
NM_001322173.1
NP_001309102.1
7, 8
938
NM_001322174.1
NP_001309103.1
7, 8
825
NM_001322175.1
NP_001309104.1
7, 8
823
NM_152906.6
NP_690870.3
8, 9
925
NR_104274.2
6, 7
544
NR_104275.2
6, 7
784
NR_136206.1
6, 7
723
NR_136211.1
8, 9
909
NR_136212.1
7, 8
805
XM_011529863.1
XP_011528165.1
8, 9
962
XM_011529865.1
XP_011528167.1
6, 7
675
XM_011529867.1
XP_011528169.1
8, 9
908
XM_017028587.1
XP_016884076.1
8, 9
1017
XM_017028588.1
XP_016884077.1
7, 8
712
GenBank mRNA
AK092484.1
-
6, 7
582
AK289862.1
-
9, 10
1028
AK295210.1
-
7, 8
939
AK297075.1
-
5, 6
430
AK297922.1
-
8, 9
769
AK298593.1
-
7, 8
656
AK301054.1
-
6, 7
503
AK309398.1
-
5, 6
424
AK316020.1
-
8, 9
772
AK316056.1
-
8, 9
1033
AL713640.1
-
5, 6
553
BC041339.1
-
8, 9
888
5284126
BF726547.1
-
4, 5
410
BM048105.1
-
394
BU173566.1
-
8, 9
844
BX345246.2
-
6, 7
738
CF552364.1
-
7, 8
784
CR456355.1
-
7, 8
740
Back To Top
More Information
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-65d5d77f49-9xzrh:80/100.66.79.6:80.
git-commit: a4c025c8ba47546866237b65de1f276a5fdbbe78
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.18.1-Offline