Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EMID1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001267895.1 | NP_001254824.1 | 15 | 1408 | ||
NM_133455.3 | NP_597712.2 | 15 | 1414 | |||
XM_005261328.2 | XP_005261385.1 | 15 | 1415 | |||
XM_005261329.2 | XP_005261386.1 | 14 | 1325 | |||
XM_011529868.2 | XP_011528170.1 | 16 | 1426 | |||
XM_011529869.2 | XP_011528171.1 | 16 | 1419 | |||
XM_011529870.2 | XP_011528172.1 | 15 | 1334 | |||
XM_011529871.2 | XP_011528173.1 | 15 | 1344 | |||
XM_011529874.2 | XP_011528176.1 | 17 | 4061 | |||
XM_011529875.2 | XP_011528177.1 | 12 | 965 | |||
XM_011529876.1 | XP_011528178.1 | 14 | 1058 | |||
XM_017028589.1 | XP_016884078.1 | 16 | 4655 | |||
GenBank mRNA | AJ416090.1 | - | 15 | 1281 | ||
BC013830.1 | - | 7 | 409 | |||
BC046358.1 | - | 15 | 1362 | 5770546 | ||
CR456407.1 | - | 15 | 1297 |