Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TYW5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039693.2 | NP_001034782.1 | 8 | 1032 | ||
NR_004862.1 | 8 | 1013 | ||||
NR_109905.1 | 5 | 761 | ||||
NR_109906.1 | 7 | 925 | ||||
NR_109907.1 | 7 | 877 | ||||
XM_017003310.1 | XP_016858799.1 | 6 | 660 | |||
GenBank mRNA | AK095272.1 | - | 7 | 641 | ||
AK293064.1 | - | 7 | 692 | |||
AK298977.1 | - | 8 | 776 | |||
BC025758.1 | - | 7 | 672 | |||
BC063502.1 | - | 4 | 301 | |||
BC132835.1 | - | 8 | 796 | 40146857 | ||
BC136837.1 | - | 8 | 796 | 9020828 | ||
BC144420.1 | - | 8 | 777 | |||
BX647677.1 | - | 8 | 1015 |