Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MITD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320417.1 | NP_001307346.1 | 5 | 758 | ||
NM_001320418.1 | NP_001307347.1 | 5 | 634 | |||
NM_001320419.1 | NP_001307348.1 | 4 | 547 | |||
NM_138798.2 | NP_620153.1 | 5 | 634 | |||
XM_011510581.2 | XP_011508883.1 | 5 | 646 | |||
XM_011510582.2 | XP_011508884.1 | 5 | 646 | |||
XM_011510583.1 | XP_011508885.1 | 5 | 732 | |||
XM_017003314.1 | XP_016858803.1 | 4 | 559 | |||
XM_017003315.1 | XP_016858804.1 | 5 | 646 | |||
GenBank mRNA | AL161992.1 | - | 3 | 2883 | ||
BC018453.1 | - | 5 | 602 | 4509694 | ||
BG504564.1 | - | 4 | 538 | |||
BU507044.1 | - | 5 | 616 |