Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MITD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320417.1 | NP_001307346.1 | 5, 6 | 789 | ||
NM_001320418.1 | NP_001307347.1 | 5, 6 | 665 | |||
NM_001320419.1 | NP_001307348.1 | 4, 5 | 578 | |||
NM_138798.2 | NP_620153.1 | 5, 6 | 665 | |||
XM_011510581.2 | XP_011508883.1 | 5, 6 | 677 | |||
XM_011510582.2 | XP_011508884.1 | 5, 6 | 677 | |||
XM_011510583.1 | XP_011508885.1 | 5, 6 | 763 | |||
XM_017003314.1 | XP_016858803.1 | 4, 5 | 590 | |||
XM_017003315.1 | XP_016858804.1 | 5, 6 | 677 | |||
GenBank mRNA | AL161992.1 | - | 3, 4 | 2914 | ||
BC018453.1 | - | 5, 6 | 633 | 4509694 | ||
BG504564.1 | - | 4, 5 | 569 | |||
BU507044.1 | - | 5, 6 | 647 |