Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MBOAT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321265.1 | NP_001308194.1 | 8, 9 | 985 | ||
NM_001321266.1 | NP_001308195.1 | 7, 8 | 907 | |||
NM_001321267.1 | NP_001308196.1 | 10, 11 | 1208 | |||
NM_138799.3 | NP_620154.2 | 9, 10 | 1131 | |||
NR_135598.1 | 9, 10 | 1131 | ||||
NR_135599.1 | 9, 10 | 1131 | ||||
NR_135600.1 | 8, 9 | 985 | ||||
NR_135601.1 | 7, 8 | 907 | ||||
NR_135602.1 | 9, 10 | 1131 | ||||
NR_135605.1 | 7, 8 | 907 | ||||
NR_135606.1 | 9, 10 | 1131 | ||||
GenBank mRNA | AB305044.1 | - | 9, 10 | 995 | ||
AK027321.2 | - | 9, 10 | 1091 | |||
AK295907.1 | - | 9, 10 | 1039 | |||
AK309818.1 | - | 7, 8 | 749 | |||
AK315881.1 | - | 8, 9 | 914 | |||
BC016005.1 | - | 6, 7 | 810 | |||
BC146871.1 | - | 9, 10 | 1075 | 9056799 | ||
BC157827.1 | - | 9, 10 | 1075 | 9057046 |