Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MBOAT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321267.1 | NP_001308196.1 | 3 | 385 | ||
NM_138799.3 | NP_620154.2 | 2 | 308 | |||
NR_135598.1 | 2 | 308 | ||||
NR_135599.1 | 2 | 308 | ||||
NR_135602.1 | 2 | 308 | ||||
NR_135603.1 | 3 | 385 | ||||
NR_135604.1 | 2 | 308 | ||||
NR_135606.1 | 2 | 308 | ||||
XM_011510313.1 | XP_011508615.1 | 2 | 268 | |||
XM_011510314.2 | XP_011508616.1 | 2 | 268 | |||
GenBank mRNA | AB305044.1 | - | 2 | 172 | ||
AK027321.2 | - | 2 | 268 | |||
AK295907.1 | - | 2 | 216 | |||
AK315881.1 | - | 1 | 91 | |||
BC146871.1 | - | 2 | 252 | 9056799 | ||
BC157827.1 | - | 2 | 252 | 9057046 | ||
BX442603.2 | - | 2 | 308 | |||
DA236556.1 | - | 2 | 221 |