Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCTD18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321547.1 | NP_001308476.1 | 5 | 799 | ||
NM_001321548.1 | NP_001308477.1 | 5 | 777 | |||
NM_001321550.1 | NP_001308479.1 | 5 | 1160 | |||
NM_152387.3 | NP_689600.2 | 5 | 1182 | |||
GenBank mRNA | AK055884.1 | - | 5 | 774 | ||
AK290867.1 | - | 5 | 773 | |||
BC045189.1 | - | 5 | 1153 | 5296647 | ||
BC059366.1 | - | 5 | 1130 | |||
BC067755.1 | - | 5 | 773 | 5299303 | ||
BU181789.1 | - | 1 | 38 |