Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCTD18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321547.1 | NP_001308476.1 | 6 | 873 | ||
NM_001321548.1 | NP_001308477.1 | 6 | 851 | |||
NM_001321550.1 | NP_001308479.1 | 6 | 1234 | |||
NM_152387.3 | NP_689600.2 | 6 | 1256 | |||
GenBank mRNA | AK290867.1 | - | 6 | 847 | ||
BC045189.1 | - | 6 | 1227 | 5296647 | ||
BC059366.1 | - | 6 | 1204 | |||
BC067755.1 | - | 6 | 847 | 5299303 | ||
BU181789.1 | - | 2 | 112 |