Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALS2CR12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127391.2 | NP_001120863.1 | 13 | 1444 | ||
NM_001289993.1 | NP_001276922.1 | 13 | 1112 | |||
NM_139163.3 | NP_631902.2 | 13 | 1513 | |||
NR_110620.1 | 12 | 1610 | ||||
XM_011510606.2 | XP_011508908.1 | 13 | 1459 | |||
XM_011510610.2 | XP_011508912.1 | 11 | 904 | |||
XM_011510612.2 | XP_011508914.1 | 7 | 611 | |||
XM_017003360.1 | XP_016858849.1 | 12 | 1096 | |||
XM_017003362.1 | XP_016858851.1 | 11 | 945 | |||
GenBank mRNA | AB053314.1 | - | 12 | 1098 | ||
AK124115.1 | - | 12 | 1556 | |||
AK292283.1 | - | 13 | 1112 | |||
BC031603.1 | - | 13 | 1424 | 5168160 | ||
BX095441.1 | - | 3 | 259 |