Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C2orf50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182500.2 | NP_872306.1 | 3 | 702 | ||
XM_006711857.2 | XP_006711920.1 | 3 | 626 | |||
XM_006711858.2 | XP_006711921.1 | 3 | 626 | |||
XM_006711859.3 | XP_006711922.1 | 3 | 626 | |||
GenBank mRNA | AK057872.1 | - | 3 | 708 | ||
AK292828.1 | - | 3 | 627 | |||
BC120930.2 | - | 3 | 669 | 40114857 |