Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBXO36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_174899.4 | NP_777559.3 | 3 | 380 | ||
XM_005246317.1 | XP_005246374.1 | 3 | 390 | |||
GenBank mRNA | AK123085.1 | - | 4 | 685 | ||
AK289655.1 | - | 3 | 365 | |||
AK309702.1 | - | 3 | 358 | |||
BC022538.1 | - | 4 | 496 | 4817261 | ||
BC033935.1 | - | 3 | 373 | 5296808 |